Merck
Functional deficit of sperm and fertility impairment in men with antisperm antibodies.
Journal of reproductive immunology (2015-09-27)
V A Bozhedomov, M A Nikolaeva, I V Ushakova, N A Lipatova, G E Bozhedomova, G T Sukhikh
ABSTRAKT
Autoimmune reactions against the sperm cells play an ambiguous role in fertility impairment. The objective of this study was to characterize functional deficit of sperm conditioned by antisperm immune response in normozoospermic men. This was a multi-centric, cross-sectional, сase-control study. The study subjects were 1060 infertile normozoospermic men and 107 fertile men. The main outcome measures were clinical examination, semen analysis including MAR test for antisperm antibodies (ASA), computer-aided sperm analysis, acrosome reaction (AR) detected with flow cytometry, DNA fragmentation measured with sperm chromatin dispersion, reactive oxygen species (ROS) assessed using the luminol-dependent chemiluminescence method. 2% of the fertile men had MAR-IgG ≥ 50%, but all subjects with MAR-IgG>12% were outliers; 16% infertile men had MAR-IgG ≥ 50% (p<0.0001). There was a direct correlation between the infertility duration and MAR-IgG (R=0.3; р<0.0001). The ASA-positive infertile men had AR disorders 2.1 times more frequently (р<0.02), predominantly inductivity disorders. We found signs of hyperactivation proportionate to the ASA level (p<0.001). DNA fragmentation was more highly expressed and was 1.6 and 1.3 times more frequent compared with the fertile and the ASA-negative patients, respectively (p<0.001 and p<0.05). We found signs of oxidative stress (OS): ROS generation by washed ASA-positive spermatozoa was 3.7 times higher than in the fertile men (p<0.00001) and depended on the ASA levels (R=0.5; p<0.0001). The ASA correlation with ROS generation in native sperm was weak (R=0.2; р<0.001). We concluded that autoimmune reactions against spermatozoa are accompanied by a fertility decrease in normozoospermia. This results from AR and capacitation disorders and DNA fragmentation. The pathogenesis of sperm abnormalities in immune infertility is associated with the OS of spermatozoa.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%