MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Vitamin C in infant formula and adult/pediatric nutritional Formula by ultra-performance liquid chromatography with Ultraviolet detection: first action 2012.22.
Vitamin C in infant formula and adult/pediatric nutritional Formula by ultra-performance liquid chromatography with Ultraviolet detection: first action 2012.22.
Journal of AOAC International (2013-11-29)
Esther Campos-Giménez, Patric Fontannaz, Karine Redeuil, Tamara Kilinc, Dawn Dowell
ABSTRACT
During the AOAC Annual Meeting held in Las Vegas, NV from September 30 to October 3, 2012, the Stakeholder Panel on Infant Formula and Adult Nutritionals convened to review single-laboratory validation data submitted for the method, Vitamin C in Adult/Pediatric Formula by Ultra-Performance Liquid Chromatography with Ultraviolet Detection. This method is a modified version of the method "HPLC-UV Determination of Total Vitamin C in a Wide Range of Fortified Food Products" previously published in Food Chem., 94, 626-631 (2006). The SLV data from the modified method were reviewed and compared to the standard method performance requirements (SMPR 2012.012), and it was concluded that the method meets the requirements. The method was approved as AOAC Official First Action. The method is based on the acidic extraction of ascorbic acid in the presence of Tris[2-carboxyethyl] phosphine (TCEP) as a reducing agent. Separation was achieved on a C18 column with a sodium acetate eluent (pH 5.4) combined with TCEP and decylamine as an ion-pairing agent. Accuracy rates were between 90 and 100%. Repeatability RSD (RSD,) ranged from 1.4 to 2.5%, and intermediate reproducibility RSD (RSDiR) ranged from 1.3 to 7.5%.
Product Description
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(+)-Sodium L-ascorbate, SAJ special grade, ≥99.0%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard