MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Vitamin C and survival among women with breast cancer: a meta-analysis.
European journal of cancer (Oxford, England : 1990) (2014-03-13)
Holly R Harris, Nicola Orsini, Alicja Wolk
ABSTRACT
The association between dietary vitamin C intake and breast cancer survival is inconsistent and few studies have specifically examined vitamin C supplement use among women with breast cancer. The purpose of this study was to summarise results from prospective studies on the association between vitamin C supplement use and dietary vitamin C intake and breast cancer-specific mortality and total mortality. Studies were identified using the PubMed database through February 6, 2014 and by examining the references of retrieved articles. Prospective studies were included if they reported relative risks (RR) with 95% confidence intervals (95% CIs) for at least two categories or as a continuous exposure. Random-effects models were used to combine study-specific results. The ten identified studies examined vitamin C supplement use (n=6) and dietary vitamin C intake (n=7) and included 17,696 breast cancer cases, 2791 total deaths, and 1558 breast cancer-specific deaths. The summary RR (95% CI) for post-diagnosis vitamin C supplement use was 0.81 (95% CI 0.72-0.91) for total mortality and 0.85 (95% CI 0.74-0.99) for breast cancer-specific mortality. The summary RR for a 100mg per day increase in dietary vitamin C intake was 0.73 (95% CI 0.59-0.89) for total mortality and 0.78 (95% CI 0.64-0.94) for breast cancer-specific mortality. Results from this meta-analysis suggest that post-diagnosis vitamin C supplement use may be associated with a reduced risk of mortality. Dietary vitamin C intake was also statistically significantly associated with a reduced risk of total mortality and breast cancer-specific mortality.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sodium ascorbate, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(+)-Sodium L-ascorbate, SAJ special grade, ≥99.0%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%