MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Spectrophotometric, spectrofluorimetric, and potentiometric assays of cetyltrimethylammonium bromide in industrial wastewater samples.
Spectrophotometric, spectrofluorimetric, and potentiometric assays of cetyltrimethylammonium bromide in industrial wastewater samples.
Journal of AOAC International (2014-08-26)
Lobna Abdel Aziz Hussein, Nermine Victor Fares, Amira Mabrouk El-Kosasy
ABSTRACT
This work deals with spectrophotometric, spectrofluorimetric, and potentiometric analyses of cetyltrimethylammonium bromide (CTAB) cationic detergent. The spectrophotometric procedure depends on measuring the absorbance of its binary complex with eosin yellow in Britton-Robinson buffer (pH 4) at Lambda max 547 nm in the range of 2.0-14.0 microg/mL with an accuracy of 100.15 +/- 0.54%. The spectrofluorimetric procedure depends on determining the quenching of the fluorescence intensity of fluorescein dye by CTAB in the presence of borate buffer at Lambda em = 500 nm, Lambda ex = 304 nm, in the range of 2.90-14.50 microg/mL with an accuracy of 99.81 + 0.33%. The electrochemical procedure describes an ionophore-based technique using a graphite sensor to measure 0.036 microg/mL and showed an accuracy of 100.11 +/- 0.61%. The experimental conditions affecting each of the three suggested procedures were studied and optimized. All the developed procedures were validated and satisfactorily applied for the determination of CTAB in industrial wastewater samples.
Product Description
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
Sigma-Aldrich
Fluorescein, free acid
Supelco
Hexadecyltrimethylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
SAFC
Hexadecyltrimethylammonium bromide, USP/NF
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, ≥99.0%, contains 200-400 ppm BHT as inhibitor, ReagentPlus®
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
Hexadecyltrimethylammonium chloride, ≥98.0% (NT)
Sigma-Aldrich
Cetyltrimethylammonium chloride solution, 25 wt. % in H2O
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Hexadecyltrimethylammonium hydroxide solution, ~25% in methanol (T)
Sigma-Aldrich
Cetyltrimethylammonium hydrogensulfate, 99%
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer