MilliporeSigma
  • Purification and characterization of cocoonase from the silkworm Bombyx mori.
Purification and characterization of cocoonase from the silkworm Bombyx mori.
Bioscience, biotechnology, and biochemistry (2014-07-19)
Hisayoshi Fukumori, Satoshi Teshiba, Yuichi Shigeoka, Kohji Yamamoto, Yutaka Banno, Yoichi Aso
ABSTRACT
Cocoonase (CCN) which facilitates the degradation of a cocoon is recognized as a trypsin-like serine protease. In this study, CCN from the silkworm Bombyx mori was purified and comprehensively characterized. Its activity was maximal at about pH 9.8. It was stable above pH 3.4 at 4 °C and below 50 °C at pH 7.5. CuSO₄, FeSO₄, and ZnSO₄ showed inhibitory effects on CCN, but other salts improved activity. Typical trypsin inhibitors inhibited CCN, but the relative inhibitory activities were much lower than those against bovine trypsin. An extract of cocoon shells inhibited trypsin, but it was only slightly inhibitory against CCN. There were significant differences in catalytic efficiencies and substrate specificities as between CCN and bovine trypsin.
Sigma-Aldrich
Acetyl chloride, reagent grade, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetyl chloride solution, 1 M in methylene chloride
Pricing and availability is not currently available.
Sigma-Aldrich
CHES, BioXtra, ≥99.0% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
CHES, ≥99.0% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Acetyl chloride, reagent grade, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Acetyl chloride, puriss. p.a., ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
CHES, BioUltra, ≥99.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Pricing and availability is not currently available.
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.