MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Cytotoxic and antimicrobial evaluations of novel apoptotic and anti-angiogenic spiro cyclic 2-oxindole derivatives of 2-amino-tetrahydroquinolin-5-one.
Cytotoxic and antimicrobial evaluations of novel apoptotic and anti-angiogenic spiro cyclic 2-oxindole derivatives of 2-amino-tetrahydroquinolin-5-one.
Archiv der Pharmazie (2015-02-11)
Said A S Ghozlan, Magda F Mohamed, Ahmed G Ahmed, Samia A Shouman, Yasmin M Attia, Ismail A Abdelhamid
ABSTRACT
A novel series of cyclic 2-oxindole derivatives incorporating 2-amino-tetrahydroquinolin-5-one were prepared. The structures of the prepared compounds were elucidated using different spectral tools. The regio-orientation of the reaction products was elucidated through NOE difference experiments and through using substituents on the ortho position to affect further cyclization. Antitumor and antimicrobial evaluations were performed on the prepared compounds. Most of these compounds exhibited high to moderate antimicrobial activity. With respect to the antitumor activity, the compounds showed more potent cytotoxic effect only toward the human breast cancer cell line MCF-7. Also, we found that derivatives containing an ester group (8c, 11b, 14b, and 15b) are more active than those containing a cyanide group (8a, 11a, 14a, and 15a). Moreover, compounds 15b and 8b are the most active derivatives in this group. These two compounds showed apoptotic inhibition of the proliferation of human breast adenocarcinoma MCF-7 cells through DNA fragmentation, induction of the tumor suppressor protein p53, induction of caspase-9, and finally the inhibition of angiogenesis by decreasing vascular endothelial growth factor expression and secretion.
Sigma-Aldrich
2-Oxindole, 97%
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
USP
Tromethamine, United States Pharmacopeia (USP) Reference Standard
SAFC
Tromethamine
Trometamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ≥99.8%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, JIS special grade, ≥99.0%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ≥99.8%