MilliporeSigma
  • Roundabout 4 regulates blood-tumor barrier permeability through the modulation of ZO-1, Occludin, and Claudin-5 expression.
Roundabout 4 regulates blood-tumor barrier permeability through the modulation of ZO-1, Occludin, and Claudin-5 expression.
Journal of neuropathology and experimental neurology (2014-12-04)
Heng Cai, Wenjing Liu, Yixue Xue, Xiuli Shang, Jing Liu, Zhen Li, Ping Wang, Libo Liu, Yi Hu, Yunhui Liu
ABSTRACT
The blood-tumor barrier (BTB) restricts the delivery of chemotherapeutic drug molecules to tumor tissues. We found that the endothelial cell (EC) receptor molecule Roundabout 4 (Robo4) is endogenously expressed in human brain microvascular ECs and that it is upregulated in a BTB model of glioma cocultured ECs. Knockdown of Robo4 in this BTB model increased permeability; short hairpin RNA targeting Robo4 (shRobo4) led to decreased transendothelial electric resistance values, increased BTB permeability, and downregulated expression of the EC tight junction proteins ZO-1, occludin, and claudin-5. Roundabout 4 influenced BTB permeability via binding with its ligand, Slit2. Short hairpin RNA targeting Robo4 also increased matrix metalloproteinase-9 (MMP-9) activity and expression in glioma cocultured ECs; pretreatment with the MMP inhibitor GM6001 partially blocked the effects of shRobo4 on the transendothelial electric resistance values and ZO-1 and occludin expression. Short hairpin RNA targeting Robo4 also upregulated the phosphorylation of Src and Erk1/2; the Src inhibitor PP2 and the Erk1/2 inhibitor PD98059 blocked shRobo4-mediated alteration in ZO-1 and occludin expression. Together, our results indicate that knockdown of Robo4 increased BTB permeability by reducing EC tight junction protein expression, and that the Src-Erk1/2-MMP-9 signal pathways are involved in this process. Thus, Robo4 may represent a useful future therapeutic target for enhancing BTB permeability.
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
(R)-N4-Hydroxy-N1-[(S)-2-(1H-indol-3-yl)-1-methylcarbamoyl-ethyl]-2-isobutyl-succinamide, >95% (HPLC)
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%