MilliporeSigma
  • Aryl hydrocarbon receptor mediates indoxyl sulfate-induced cellular senescence in human umbilical vein endothelial cells.
Aryl hydrocarbon receptor mediates indoxyl sulfate-induced cellular senescence in human umbilical vein endothelial cells.
Journal of atherosclerosis and thrombosis (2014-04-15)
Masayuki Koizumi, Junko Tatebe, Ippei Watanabe, Junichi Yamazaki, Takanori Ikeda, Toshisuke Morita
ABSTRACT
Vascular senescence, which is accelerated in individuals with chronic kidney disease (CKD), contributes to the development of cardio-renal syndrome, and various uremic toxins may play important roles in the mechanisms underlying this phenomenon. We recently reported that indoxyl sulfate (IS), a uremic toxin, directly activates aryl hydrocarbon receptor (AhR) and generates oxidative stress through NADPH oxidase-4 in human umbilical vein endothelial cells (HUVECs). In the current study, we sought to examine whether IS regulates sirtuin 1 (Sirt1) and affects endothelial senescence via AhR activation. HUVECs were incubated with 500 μmol/L of IS for the indicated time periods. In order to evaluate changes in the senescence of the HUVECs, the number of senescence-associated β-galactosidase (SA β-gal)-positive cells was determined using an image analysis software program. The intracellular nicotinamide phosphoribosyltransferase (iNampt) activity, cellular NAD(+)/NADPH ratio and Sirt1 activity were analyzed according to a colorimetric assay to determine the mechanism of cellular senescence. Furthermore, we evaluated the involvement of AhR in the senescence-related changes induced by IS using AhR antagonists. IS decreased the iNampt activity, NAD(+)/NADPH ratio and Sirt1 activity, resulting in an increase in the percentage of SA β-gal-positive cells. On the other hand, the AhR antagonists restored the IS-induced decrease in the NAD(+) content in association with an improvement in the iNampt activity and ameliorated the senescence-related changes. Taken together, these results indicate that IS impairs the iNampt-NAD(+)-Sirt1 system via AhR activation, which in turn promotes endothelial senescence. The IS-AhR pathway induces endothelial senescence. Therefore, blocking the effects of AhR in the endothelium may provide a new therapeutic tool for treating cardio-renal syndrome.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Pricing and availability is not currently available.
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Pricing and availability is not currently available.
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Pricing and availability is not currently available.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Pricing and availability is not currently available.
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Pricing and availability is not currently available.
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Acetovanillone, ≥98%, FG
Pricing and availability is not currently available.