MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • LDL-R promoting activity of peptides derived from human PCSK9 catalytic domain (153-421): design, synthesis and biochemical evaluation.
LDL-R promoting activity of peptides derived from human PCSK9 catalytic domain (153-421): design, synthesis and biochemical evaluation.
European journal of medicinal chemistry (2015-02-14)
Rasha H Alghamdi, Paul O'Reilly, Chunyu Lu, James Gomes, Thomas A Lagace, Ajoy Basak
ABSTRACT
High level of Low Density Lipoprotein-Cholesterol (LDL-C) in circulation in the blood is associated with an elevated risk of cardiovascular disease (CVD) and stroke. Currently the statin drugs which inhibit the enzyme HMG-CoA reductase responsible for cholesterol synthesis in the liver are very effective in lowering LDL-cholesterol. However these drugs are often associated with serious side effects particularly for ∼10-12% of cases. Therefore there is a need to develop non-statin based cholesterol reducing agents. Recently it was revealed that the secreted Proprotein Convertase Subtilisin Kexin 9 (PCSK9) binds with LDL-receptor (LDL-R) causing its degradation in the lysosome with the result of LDL-C accumulating in the blood. Thus PCSK9 has become an alternative target for development of non-statin cholesterol reducing agents. It is established that the catalytic domain of PCSK9 (aa153-421) and the EGF-A domain of LDL-R (aa314-355) are involved in the above bind leading to the reduction of LDL-R level and accumulation of LDL-C. The major goal of this study is to identify peptide/s from the catalytic domain of hPCSK9 that can block the binding of hPCSK9 and LDL-R and therefore can reduce LDL-R degradation leading to the clearance of LDL-C from the plasma. Using 51 synthetic linear peptides (P1-P51) of 15aa long with 10 amino acids overlapping sequences spanning the entire catalytic segment of hPCSK9 (aa153-421), we identified two domains of hPCSK9 namely (aa323-358) and (aa365-384) that exhibited strong binding affinity towards synthetic EGF-A peptide. The results were based on mass spectrometry, fluorescence spectroscopy and native gel electrophoresis. Thus peptides containing the above segments in part (P35-P39 and P42-P47) exhibited LDL-R promoting activity when added exogenously to culture medium of growing human hepatic cells like HepG2 and HuH7. The effects were particularly significant with peptides P36, P37, P46 and P47. Interestingly, the first two peptides are present within the disulphide loop Cys(323)-Cys(358) and contain the key gain of function mutation D(374)/Y site while the last two peptides contain another disulphide bridge loop Cys(375)-Cys(378) and the second most potent gain of function mutation R(357)/H. Further studies revealed that S-S bridged cyclic loop peptide hPCSK9(365-384) exhibited the highest (∼3.5-fold) LDL-R promoting activity in both HepG2 and HuH7 when applied at 5 μM concentration level. This effect is completely abrogated when one of the Cys residues is substituted by Ala thereby preventing any S-S bond formation. This suggested its critical role in the bioactivity. It is proposed that LDL-R promoting activity of this and other selected PCSK9 catalytic peptides such as P36, P37, P46 and P47 are most likely mediated via intervention of PCSK9:LDL-R complex formation. Our findings may find useful application in future development of small molecule PCSK9 inhibitors for intervention of hypercholesterolemia and associated cardiovascular disease.
Product Description
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Supelco
Sinapic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, SAJ special grade, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)