MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Assessment of endosulfan induced genotoxicity and mutagenicity manifested by oxidative stress pathways in freshwater cyprinid fish crucian carp (Carassius carassius L.).
Assessment of endosulfan induced genotoxicity and mutagenicity manifested by oxidative stress pathways in freshwater cyprinid fish crucian carp (Carassius carassius L.).
Chemosphere (2014-08-17)
Sabzar Ahmad Dar, Abdul Rehman Yousuf, Masood-ul-Hassan Balkhi, Farooq Ahmad Ganai, Farooz Ahmad Bhat
ABSTRACT
Over the past few decades, endosulfan, one of the polychlorinated pesticides still in use, has received considerable attention of a number of international regulations and restriction action plans worldwide. This study aimed to evaluate the cytogenetic effects of endosulfan using robust genotoxicity assays, along with the oxidative stress pathways in order to understand biochemical mechanism, in Carassius carassius L. The LC50-96 h (95% confidence limits) value of endosulfan was 0.070 (0.046-0.093) ppm; and on its basis three test concentrations (sub-lethal I: 0.052, II: 0.035 and III: 0.017 ppm) were selected for 35 d in vivo exposure. The mean concentration of endosulfan in aquaria was always constant, when analyzed by dispersive liquid-liquid micro extraction (DLLME) followed by GC-MS. Autopsy was done on days 1, 2, 3, 4, 7, 14, 21, 28 and 35 of endosulfan exposure; the micronucleus formation (MN), authenticated by scanning electron microscopy, and chromosomal aberrations (CA), were induced significantly (p<0.05) in all the treated groups, including positive control cyclophosphamide (4 ppm), when compared to negative control. Similarly lipid peroxidation (LPO) was induced significantly with the maximal at higher concentration (SL-I) on 4th day (722.45%; p<0.01). Antioxidant biomarkers like glutathione reduced, superoxide dismutase and catalase also fluctuated significantly (p<0.01) in all treatment groups. Collective findings demonstrated that genotoxic effects were invariably accompanied and correlated with increased oxidative stress and disturbance of antioxidant enzymes; and the MN and CA assays are useful tools in determining potential genotoxicity of aquatic xenobiotics and might be appropriate as a part of monitoring program.
Product Description
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Potassium periodate, 99.8%, ACS reagent
Sigma-Aldrich
Pyrogallol, ACS reagent
Sigma-Aldrich
Pyrogallol, ACS reagent, ≥99%
Sigma-Aldrich
Pyrogallol, ≥98% (HPLC)
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Pyrogallol, JIS special grade, ≥99.0%
Supelco
Pyrogallol, analytical standard
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Cyclophosphamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Supelco
Endosulfan, PESTANAL®, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder