MilliporeSigma
  • Metabolism of palmatine by human hepatocytes and recombinant cytochromes P450.
Metabolism of palmatine by human hepatocytes and recombinant cytochromes P450.
Journal of pharmaceutical and biomedical analysis (2014-10-07)
Jiri Vrba, Barbora Papouskova, Michaela Pyszkova, Martina Zatloukalova, Karel Lemr, Jitka Ulrichova, Jan Vacek
ABSTRACT
In this study, we developed a new liquid chromatography-mass spectrometry (LC-MS) method for analysis of the protoberberine alkaloid palmatine and its metabolites with separation performed on a cyanopropyl-modified stationary phase. Palmatine (10 μM) was metabolized using suspensions of human hepatocytes and human recombinant cytochrome P450 (CYP) enzymes. Our analyses using electrospray ionization-quadrupole time-of-flight mass spectrometry revealed that palmatine was relatively resistant to the metabolic activity of human hepatocytes and recombinant CYP enzymes. However, we found that the biotransformation of palmatine in human hepatocytes included O-demethylation or hydroxylation, and that the product of palmatine demethylation was conjugated by glucuronidation or sulfation. Moreover, we found that human recombinant CYP2D6 and, to a lesser extent, CYP1A2 can mediate O-demethylation of palmatine. These results provide fundamental insights into the biotransformation of palmatine in human in vitro models and, together with the LC-MS method, can be applied for further studies on the biotransformation of palmatine and other protoberberine alkaloids.
Product Description
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard