MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Targeting mTOR and AREG with everolimus, sunitinib and sorafenib in HPV-positive and -negative SCC.
Targeting mTOR and AREG with everolimus, sunitinib and sorafenib in HPV-positive and -negative SCC.
Anticancer research (2015-04-12)
Christoph Aderhold, Anne Faber, Claudia Umbreit, Richard Birk, Christel Weiss, Jörg Ulrich Sommer, Karl Hörmann, Johannes David Schultz
ABSTRACT
Head and neck squamous cell carcinoma (HNSCC) is an aggressive epithelial malignancy. It is the most common neoplasm appearing in the upper aerodigestive tract and the sixth most common cancer worldwide. The five-year survival rate remains poor despite advances in surgery, radiation and chemotherapy. Furthermore, the incidence of human papillomavirus (HPV)-associated oropharyngeal cancer is rising. Thus, innovative therapy approaches are imperative in order to improve the situation. Everolimus, an inhibitor of the mammalian target of rapamycin (mTOR) and sorafenib and sunitinib, multityrosine kinase inhibitors, have been notably effective in the therapy of different tumor entities. The modest side-effects and oral application of the drugs might improve patient compliance. Expression levels of mTOR and Amphiregulin (AREG) in p16-positive and -negative SCC (squamous cell carcinoma) and the effect of everolimus, sorafenib or sunitinib on the expression levels of these target proteins were assessed. As far as we are aware of, this is one of the first in vitro studies to evaluate the effect of these small-molecule drugs with regard to the p16 status of SCC cells. p16-negative HNSCC 11A and 14C cells and p16-positive CERV196 cells were exposed to different concentrations of everolimus, sorafenib and sunitinib for 2-8 days. Expression levels of mTOR and AREG were determined by enzyme-linked immunosorbent assay (ELISA) and compared against a chemonaïve control. AREG and mTOR were expressed in all tested cell lines. CERV196 displayed a remarkable increase of mTOR expression compared to p16-negative HNSCC. On the contrary, AREG levels were reduced by 50% in CERV196. Everolimus, sorafenib and sunitinib significantly reduced mTOR expression. Everolimus significantly decreased AREG expression independently of the HPV status. Sunitinib and sorafenib increased AREG expression in HNSCC 11A and 14C but not in CERV196. The applied drugs showed remarkable suppression of mTOR expression, which might delay tumor progression. Interestingly, sorafenib and sunitinib increased AREG in HNSCC 11A and 14C, which could be a possible evasive mechanism following incubation with these drugs. On the contrary, p16-positive CERV196 showed increased susceptibility to sorafenib and sunitinib concerning suppression of AREG expression. Further studies are required to evaluate the HPV-dependent differences of therapy response and the possible consequences for treatment options.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%