MilliporeSigma
  • Direct role of antibody-secreting B cells in the severity of chronic hepatitis B.
Direct role of antibody-secreting B cells in the severity of chronic hepatitis B.
Journal of medical virology (2014-08-29)
Ayaid Khadem Zgair, Jenan A Ghafil, Razaq H E Al-Sayidi
ABSTRACT
Chronic hepatitis B involves different immune cells. The direct role of antibody-secreting B cells in the severity of chronic hepatitis B unclear. In this study, the number of plaque forming cells [PFC-(IgG, IgM, anti-HBc IgG, and anti-HBc IgM)], liver function tests (LFT) [alkaline phosphatase (ALP), alanine aminotransferase (ALT), and total serum bilirubin (TSB)], the levels of IL-10 in sera and in lymphocyte cultures, the number of CD4(+) and CD8(+) cells were measured in the peripheral blood of patients and in the controls. In addition, the hepatocytotoxic effect of anti-HBc and anti-HBe in vitro was studied. The largest number of PFCs was observed in the peripheral blood of patients with chronic hepatitis B. This was concomitant with a decrease in CD4(+) /CD8(+) ratio versus this ratio in asymptomatic HBV carriers and in healthy volunteers (P < 0.05). An increase in immunoglobulin (IgG and IgM) levels, anti-HBc IgG, and anti-HBc IgM levels and LFTs in peripheral blood of patients with chronic hepatitis B was seen. Anti-HBc induced hepatocytotoxicity in vitro. The expression of mRNA and protein for IL-10 production was observed at a significant level in culture of lymphocytes isolated from patients with chronic hepatitis B. In addition, a high level of IL-10 was found only in the sera of patients with chronic hepatitis B. It is concluded that the antibody-secreting B cells and the antibodies, which are produced, play an important role in the severity of chronic hepatitis B, which was related negatively with CD4(+) /CD8(+) ratio and positively with IL-10 expression.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.