MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Cytotoxic effect of the immunotoxin constructed of the ribosome-inactivating protein curcin and the monoclonal antibody against Her2 receptor on tumor cells.
Cytotoxic effect of the immunotoxin constructed of the ribosome-inactivating protein curcin and the monoclonal antibody against Her2 receptor on tumor cells.
Bioscience, biotechnology, and biochemistry (2015-02-24)
Lidia Patricia Jaramillo-Quintero, Arturo Contis Montes de Oca, Andrés Romero Rojas, Saúl Rojas-Hernández, Rafael Campos-Rodríguez, Alma Leticia Martínez-Ayala
ABSTRACT
The toxicity of the curcin on cancer cells allows to consider this protein as the toxic component of an immunotoxin directed to Her2, which is associated with cancer. Reductive amination was proposed to conjugate curcin and an anti-Her2; the binding was tested using Polyacrylamide gel electrophoresis, western blot, and immunocytochemistry. The in vitro cytotoxicity of curcin and the immunotoxin was assessed on breast cancer cell lines SK-BR-3 (Her2(+)) and MDA-MB-231 (Her2(-)). IC50 values for curcin were 15.5 ± 8.3 and 18.6 ± 2.4 μg/mL, respectively, statistically equivalent (p < 0.05). While to the immunotoxin was 2.2 ± 0.08 for SK-BR-3 and 147.6 ± 2.5 μg/mL for MDA-MB-231. These values showed that the immunotoxin was seven times more toxic to the SK-BR-3 than curcin and eight times less toxic to the MDA-MB-231. The immunotoxin composed of curcin and an antibody against Her2 and constructed by reductive amination could be a therapeutic candidate against Her2(+) cancer.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water
Sigma-Aldrich
4-Chloro-1-naphthol, crystalline
Sigma-Aldrich
4-Chloro-1-naphthol, tablet
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium (meta)periodate, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Sodium cyanoborohydride, reagent grade, 95%
Sigma-Aldrich
Sodium cyanoborohydride solution, 1.0 M in THF
Sigma-Aldrich
Sodium cyanoborohydride solution, 5.0 M in 1 M NaOH