MilliporeSigma
  • Novel spectrophotometric method for detection and estimation of butanol in acetone-butanol-ethanol fermenter.
Novel spectrophotometric method for detection and estimation of butanol in acetone-butanol-ethanol fermenter.
Talanta (2015-05-13)
Sampa Maiti, Saurabh Jyoti Sarma, Satinder Kaur Brar, Yann Le Bihan, Patrick Drogui, Gerardo Buelna, Mausam Verma, Carlos Ricardo Soccol
ABSTRACT
A new, simple, rapid and selective spectrophotometric method has been developed for detection and estimation of butanol in fermentation broth. The red colored compound, produced during reduction of diquat-dibromide-monohydrate with 2-mercaptoethanol in aqueous solution at high pH (>13), becomes purple on phase transfer to butanol and gives distinct absorption at λ520nm. Estimation of butanol in the fermentation broth has been performed by salting out extraction (SOE) using saturated K3PO4 solution at high pH (>13) followed by absorbance measurement using diquat reagent. Compatibility and optimization of diquat reagent concentration for detection and estimation of butanol concentration in the fermentation broth range was verified by central composite design. A standard curve was constructed to estimate butanol in acetone-ethanol-butanol (ABE) mixture under optimized conditions. The spectrophotometric results for butanol estimation, was found to have 87.5% concordance with the data from gas chromatographic analysis.
Sigma-Aldrich
Butyric acid, SAJ special grade, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for absorption spectrum analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, suitable for residue analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, JIS 1000, ≥99.5%, suitable for residue analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium phosphate monobasic, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Pricing and availability is not currently available.
Sigma-Aldrich
Ethanol, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
Butyric acid, natural, ≥99%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Butyric acid, ≥99%, FG
Pricing and availability is not currently available.
Sigma-Aldrich
Butyric acid, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, suitable for luminescence, Molecular Biology, BioUltra, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Pricing and availability is not currently available.