MilliporeSigma
  • The protein tyrosine phosphatase DEP-1/PTPRJ promotes breast cancer cell invasion and metastasis.
The protein tyrosine phosphatase DEP-1/PTPRJ promotes breast cancer cell invasion and metastasis.
Oncogene (2015-03-17)
K Spring, P Fournier, L Lapointe, C Chabot, J Roussy, S Pommey, J Stagg, I Royal
ABSTRACT
DEP-1/PTPRJ is a receptor-like protein tyrosine phosphatase mainly known for its antiproliferative and tumor-suppressive functions. Many identified substrates are growth factor receptors, and DEP-1 is deleted and/or mutated in human cancers including that of the breast. However, DEP-1 was also identified as a promoter of Src activation and proinvasive functions in the endothelium, suggesting it could perhaps mediate breast cancer invasiveness that is likewise driven by Src family kinases. We show here that DEP-1 expression was greater in highly invasive breast cancer cells (MDA-MB-231, Hs578T, BT-549) than in the less invasive or untransformed cell lines tested (MCF-7, T47D, SK-BR3 and MCF10A). DEP-1 silencing experiments in invasive cells demonstrated that moderately expressed and catalytically active DEP-1 was required, in collaboration with basal epidermal growth factor receptor activity, for Src activation and the phosphorylation of its substrate Cortactin, and for their colocalization at the cell's leading edge. This correlated with an increased number of cell protrusions, and an enhanced capacity of the cells to migrate and invade. Similarly, moderate overexpression of DEP-1 in the low-invasive cells resulted in the promotion of their invasiveness in an Src-dependent manner. Consistent with these data, the expression of endogenous DEP-1 was elevated in a bone metastatic cell line derived from MDA-MB-231 cells, and promoted increased Src Y418 and Cortactin Y421 phosphorylation, as well as pro-MMP9 secretion and Matrigel invasion. Importantly, the silencing of DEP-1 in MDA-MB-231 cells greatly decreased their ability to metastasize, despite having no effect on tumor growth or angiogenesis. Hence, we found that moderate expression of DEP-1 was associated with the increased relapse and decreased survival of breast cancer patients. These results therefore identify a new and unsuspected role for DEP-1 as a mediator of an invasive cell program implicating Src activation and the promotion of breast cancer progression.
Sigma-Aldrich
MISSION® esiRNA, targeting human PTPRJ
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
PP2, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ptprj
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%