MilliporeSigma
miR-133a inhibits cervical cancer growth by targeting EGFR.
Oncology reports (2015-07-03)
Xuesong Song, Bo Shi, Kexin Huang, Wenjie Zhang
ABSTRACT
MicroRNAs (miRNAs) are small non-coding RNAs that play critical roles in cervical carcinogenesis and progression. microRNA-133a (miR-133a) has been reported to play a tumor-suppressor role in a range of cancers. However, the role and underlying molecular mechanism of miR-133a in cervical cancer have not been investigated. In the present study, we investigated the role of miR-133a in the tumorigenicity of cervical cancer cells in vivo and in vitro. The expression of miR-133a was investigated using real-time reverse transcription-polymerase chain reaction (qRT-PCR) in 30 cervical specimens and matched adjacent normal tissues and cervical cancer cell lines. We found that the expression level of miR‑133a was significantly downregulated in cervical cancer tissues and cervical cancer cell lines, and the aberrant expression of miR-133a was correlated with lymph node metastasis, histological grade and FIGO stage. The role of miR-133a in tumorigenicity of cervical cancer cells was assessed by the restoration of miR-133a. We found that restoration of miR‑133a inhibited cell proliferation, colony formation, migration and invasion, promoted cell apoptosis in vitro and suppressed tumorigenicity in vivo. The epidermal growth factor receptor (EGFR) was confirmed to be a direct target of miR-133a in cervical cancer cells using luciferase assay and western blotting. Restoration of miR-133a inhibited EGFR expression and activated the AKT and ERK signaling pathways. These results showed that miR-133a suppresses cervical cancer growth in vitro and in vivo through targeting EGFR, suggesting that miR-133a can be a potential target for the treatment of cervical cancer.
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Akt1 active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥75% (SDS-PAGE)
Sigma-Aldrich
MNK1 active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE)
Sigma-Aldrich
Dimethyl sulfide, ≥99%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfide, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfide, anhydrous, ≥99.0%
Sigma-Aldrich
MISSION® esiRNA, targeting human AKT1, RP11-982M15.2
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Ethanol, ≥99.5%, SAJ super special grade
Sigma-Aldrich
Ethanol, JIS 300, ≥99.5%, suitable for residue analysis
Sigma-Aldrich
Ethanol, ≥99.5%
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for fluorescence
Sigma-Aldrich
Ethanol, ≥99.5%, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfide, natural, ≥99%, FCC, FG
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%