Merck
Unsaturated guluronate oligosaccharide enhances the antibacterial activities of macrophages.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2014-03-07)
Xu Xu, Decheng Bi, Xiaoting Wu, Qingqing Wang, Gaobin Wei, Lianli Chi, Zedong Jiang, Tatsuya Oda, Min Wan
ABSTRAKT
Alginate from marine seaweeds is receiving continuous attention owing to its wide physiological activities. Herein, we sought to elucidate possible effects of alginate-derived polyguluronate (PG) and unsaturated guluronate oligosaccharide (GOS) on antibacterial activities of macrophages. Our results showed that, in contrast to PG, GOS markedly increased the phagocytosis of IgG-opsonized Escherichia coli and Staphylococcus aureus and further inhibited the survival of intracellular bacteria in macrophages. In line with this, GOS treatment resulted in the enhanced expression of Fcγ receptors on macrophages. In addition, GOS activated NF-κB pathway, induced TNF-α secretion, and elevated the expression of inducible nitric oxide synthase and the production of nitric oxide. Meanwhile, GOS stimulated the production of reactive oxygen species in macrophages. Moreover, guluronate trimer to hexamer (G3-G6) in GOS exhibited significant activity that increased the bacterial phagocytosis of macrophages, with the pentamer (G5), displaying the highest activity. Finally, our in vivo results further confirmed that GOS but not PG significantly improved bacterial clearance in murine acute peritonitis. In conclusion, GOS enhances antibacterial activities of macrophages via modulating signaling pathways related to innate immunity, suggesting that GOS might be a promising therapeutic candidate to improve the host defense against bacterial infection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O