Merck
Modulation of the cancer cell transcriptome by culture media formulations and cell density.
International journal of oncology (2015-03-18)
Seung Wook Kim, Sun-Jin Kim, Robert R Langley, Isaiah J Fidler
ABSTRAKT
We investigated how varying the composition of cell culture formulations and growing cancer cells at different densities might affect tumor cell genotype. Specifically, we compared gene expression profiles generated by human MDA‑MB‑231 breast cancer cells cultured in different media [minimum essential medium (MEM), Dulbecco's modified Eagle's medium (DMEM), or Roswell Park Memorial Institute (RPMI)‑1640 medium] containing different concentrations of fetal bovine serum (FBS) or different sera (equine or bovine) that were grown at different cell densities. More than 2,000 genes were differentially modulated by at least a 2‑fold difference when MDA‑MB‑231 cancer cells were 90% confluent and compared with cultures that were 50% confluent. Altering the concentration of serum produced an even more pronounced effect on MDA‑MB‑231 cancer cell gene expression in that 2,981 genes were differentially expressed in a comparison between cells cultured in 0.1% FBS and same cell density cultures that were maintained in 10% FBS. A comparison between MDA‑MB‑231 cancer cells that were 90% confluent in MEM, DMEM, or RPMI‑1640 media, all containing 10% FBS, resulted in 8,925 differentially expressed genes. Moreover, one‑quarter (25.6%) of genes from our genome‑wide expression analysis were expressed at significantly different levels by cells grown in MEM, DMEM, or RPMI‑1640 media. Genes associated with epithelial‑mes-enchymal transition (EMT) were among the genes that were differentially modulated by cells grown in different cell culture formulations and these genes were verified at the protein level. Collectively, these results underscore the importance of accurate reporting and maintenance of uniform culture conditions to ensure reproducible results.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O