Merck
  • Bone morphogenic protein-7 contributes to cerebral ischemic preconditioning induced-ischemic tolerance by activating p38 mitogen-activated protein kinase signaling pathway.
Bone morphogenic protein-7 contributes to cerebral ischemic preconditioning induced-ischemic tolerance by activating p38 mitogen-activated protein kinase signaling pathway.
Inflammation (2014-04-01)
Junhong Guan, Han Li, Tao Lv, Duo Chen, Ye Yuan, Shengtao Qu
ABSTRAKT
Cerebral ischemic preconditioning (IPC), which refers to a transient and noninjurious ischemia is able to induce tolerance against the subsequent lethal ischemia, including ischemic stroke. We have previously reported that bone morphogenic protein-7 (BMP-7) contributes to the neuroprotective effects of IPC-induced ischemic tolerance, and thus ameliorates the following ischemia/reperfusion (I/R) injury in rats. Consequently, in the present study, we continued to explore the underlying regulatory mechanisms involved in BMP-7-mediated cerebral IPC in the rat model of ischemic tolerance. Male Wistar rats were preconditioned by 15-min middle cerebral artery occlusion (MCAO). After 2-day reperfusion, these animals were subjected to prolonged MCAO for 2 h. Our results showed that the phosphorylated p38 mitogen-activated protein kinase (MAPK) paralleling to BMP-7 was up-regulated by IPC in rat brain. Inactivation of p38 MAPK by pretreatment of SB203580, a p38 MAPK-specific suppressor, weakened the protective effect of IPC on CA1 neurons. Moreover, the enhanced phosphorylation of p38 MAPK induced by IPC was attenuated when the endogenous BMP-7 was inhibited by BMP-7 antagonist noggin. Besides, blockade of p38 MAPK signal transduction pathway via SB203580 abrogated the protective effects of exogenous BMP-7 against cerebral infraction. These present findings suggest that BMP-7 contributes to cerebral IPC-induced ischemic tolerance via activating p38 MAPK signaling pathway.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard