Merck
Developing osteoblasts as an endpoint for the mouse embryonic stem cell test.
Reproductive toxicology (Elmsford, N.Y.) (2015-05-02)
Xinrong Chen, Deborah K Hansen, Gwenn Merry, Christian DeJarnette, Greg Nolen, Daniel Sloper, J Edward Fisher, Wafa Harrouk, Melissa S Tassinari, Amy L Inselman
ABSTRAKT
The mouse Embryonic Stem cell Test (EST) using cardiomyocyte differentiation is a promising in vitro assay for detecting potential embryotoxicity; however, the addition of another differentiation endpoint, such as osteoblasts, may improve the predictive value of the test. A number of variables such as culture conditions and starting cell number were investigated. A 14 day direct plating method of D3 mouse embryonic stem cells (mESCs) was used to test the predictivity of osteoblast differentiation as an endpoint in the EST. Twelve compounds were tested using the prediction model developed in the ECVAM validation study. Eight of the compounds selected from the EST validation study served as model compounds; four additional compounds known to produce skeletal defects were also tested. Our results indicate comparable chemical classification between the validated cardiomyocyte endpoint and the osteoblast endpoint. These results suggest that differentiation to osteoblasts may provide confirmatory information in predicting embryotoxicity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DPH, ≥98% (HPLC)
SAFC
Methotrexate
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Methotrexate hydrate, powder, BioReagent, suitable for cell culture, ≥98% (HPLC)
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Diphenhydramine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Hydroxyurea, 98%, powder
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)