Merck
Intracellular ROS Generation Mediates Maleimide-induced Cytotoxicity in Leukemia Cells.
Anti-cancer agents in medicinal chemistry (2015-06-30)
Daiane Rosolen, Vania F Noldin, Evelyn Winter, Fabiola B Filippin-Monteiro, Fatima Campos-Buzzi, Valdir Cechinel-Filho, Tania B Creczynski-Pasa
ABSTRAKT
Maleimides consist of an important class of compounds easily synthesized with multiple functional group modification that provides expressive pharmacological properties including, antitumoral activity, mediated mainly by oxidative stress. For this reason, the present study was designed to evaluate the cytotoxicity and the role of reactive oxygen species (ROS) in maleimide-induced cell death. Cell viability assays were performed to determine the cell death type in leukemia cell line induced by the compounds. The oxidative stress in maleimidetreated cells was characterized by antioxidant enzymes activities, intracellular ROS generation, and lipid peroxidation. In addition, we evaluated mitochondrial membrane potential and ATP level. Maleimide derivatives exhibited cytotoxic effects in leukemia cells with significantly increased ROS generation. However, cell viability was partly restored by catalase-treated cells. Caspases activities and caspase-independent key genes related to apoptosis were not altered by maleimides, suggesting necrosis as the main cell death process. Maleimide-induced necrosis was associated with oxidative stress, as an imbalance between ROS levels and glutathione reductase (GR) activity. This damage was also demonstrated by loss of mitochondrial membrane potential (MMP) and ATP depletion in cells treated with maleimide derivatives. These findings strongly confirmed that maleimide derivatives promoted cell death in leukemia cells triggered by oxidative stress, indicating that these compounds might be promising antitumor agents or lead molecules.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidium bromide solution, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Ethidium bromide solution, suitable for fluorescence, ~1% in H2O
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (HPLC), powder
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethidium bromide, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Propidium iodide solution