Merck
  • Mechanistic Evidence of Viscum schimperi (Viscaceae) Antihyperglycemic Activity: From a Bioactivity-guided Approach to Comprehensive Metabolite Profiling.
Mechanistic Evidence of Viscum schimperi (Viscaceae) Antihyperglycemic Activity: From a Bioactivity-guided Approach to Comprehensive Metabolite Profiling.
Phytotherapy research : PTR (2015-08-04)
Hossam M Abdallah, Mohamed A Farag, Ashraf B Abdel-Naim, Salah A Ghareib, Essam A Abdel-Sattar
ABSTRAKT
Diabetes mellitus is possibly the world's largest growing metabolic disorder. Effective treatment of diabetes is increasingly dependent on active constituents of medicinal plants capable of controlling hyperglycemia as well as its secondary complications. Viscum schimperi Engl. is a plant growing in Saudi Arabia and known for its antidiabetic activity. The potential antidiabetic activity of its methanol extract as well as its chloroform, n-butanol, and the remaining water fractions was evaluated in streptozotocin-induced diabetic rats at two dose levels. The antidiabetic activity was assessed through the determination of fasting blood glucose level, insulin levels, area under the curve (AUC) in oral glucose tolerance test, glucose absorption in isolated rat gut assay, and glucose uptake by psoas muscle. Moreover, large-scale untargeted metabolite profiling of methanol extract was performed via UPLC-PDA and qTOF-MS (ultra-performance liquid chromatography photodiode array detection and quadrupole time-of-flight mass spectrometry) respectively, to explore its chemical composition and standardization of its extract. Multivariate statistical analysis including principal component analysis and orthogonal projection to latent structures discriminant analysis was used to determine bioactives in its fractions. In conclusion, oleanane triterpenes and O-caffeoyl quinic acid conjugates were the major compounds that might account for antihyperglycemic effect of the plant.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Gliclazide, powder, ≥98%
Sigma-Aldrich
Umbelliferone, suitable for fluorescence indicator, ≥98.0% (HPLC)
Sigma-Aldrich
Umbelliferone, 99%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Naringenin, natural (US), 98%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Ammonium molybdate, 99.98% trace metals basis
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
(±)-Naringenin, ≥95%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Ammonium molybdate tetrahydrate, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Ammonium molybdate, meets USP testing specifications
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)