MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Cytoskeletal regulatory gene expression and migratory properties of B-cell progenitors are affected by the ETV6-RUNX1 rearrangement.
Cytoskeletal regulatory gene expression and migratory properties of B-cell progenitors are affected by the ETV6-RUNX1 rearrangement.
Molecular cancer research : MCR (2014-07-26)
Chiara Palmi, Grazia Fazio, Angela M Savino, Julia Procter, Louise Howell, Valeria Cazzaniga, Margherita Vieri, Giulia Longinotti, Ilaria Brunati, Valentina Andrè, Pamela Della Mina, Antonello Villa, Mel Greaves, Andrea Biondi, Giovanna D'Amico, Anthony Ford, Giovanni Cazzaniga
ABSTRACT
Although the ETV6-RUNX1 fusion is a frequent initiating event in childhood leukemia, its role in leukemogenesis is only partly understood. The main impact of the fusion itself is to generate and sustain a clone of clinically silent preleukemic B-cell progenitors (BCP). Additional oncogenic hits, occurring even several years later, are required for overt disease. The understanding of the features and interactions of ETV6-RUNX1-positive cells during this "latency" period may explain how these silent cells can persist and whether they could be prone to additional genetic changes. In this study, two in vitro murine models were used to investigate whether ETV6-RUNX1 alters the cellular adhesion and migration properties of BCP. ETV6-RUNX1-expressing cells showed a significant defect in the chemotactic response to CXCL12, caused by a block in CXCR4 signaling, as demonstrated by inhibition of CXCL12-associated calcium flux and lack of ERK phosphorylation. Moreover, the induction of ETV6-RUNX1 caused changes in the expression of cell-surface adhesion molecules. The expression of genes regulating the cytoskeleton was also affected, resulting in a block of CDC42 signaling. The abnormalities described here could alter the interaction of ETV6-RUNX1 preleukemic BCP with the microenvironment and contribute to the pathogenesis of the disease. Alterations in the expression of cytoskeletal regulatory genes and migration properties of BCP represent early events in the evolution of the disease, from the preleukemic phase to the clinical onset, and suggest new strategies for effective eradication of leukemia.
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, SAJ special grade, ≥95.0%
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥90.0% (HPLC)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Mifepristone, ≥98%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O