MilliporeSigma
  • Involvement of L-arginine/NO/cGMP/K(ATP) channel pathway in the peripheral antinociceptive actions of ellagic acid in the rat formalin test.
Involvement of L-arginine/NO/cGMP/K(ATP) channel pathway in the peripheral antinociceptive actions of ellagic acid in the rat formalin test.
Pharmacology, biochemistry, and behavior (2014-10-04)
Behnam Ghorbanzadeh, Mohammad Taghi Mansouri, Ali Asghar Hemmati, Bahareh Naghizadeh, Seyyed Ali Mard, Anahita Rezaie
ABSTRACT
The present study was conducted to evaluate the local antinociceptive actions of EA and the possible involvement of l-arginine/NO/cGMP/KATP channel pathway in this effect using formalin test in rats. To evaluate the involvement of l-arginine/NO/cGMP/KATP channel pathway in the antinociceptive action of EA, rats were pre-treated intraplantarlly with l-NAME (NOS inhibitor, 25-100μg/paw), methylene blue (guanylyl cyclase inhibitor, 100-400μg/paw), glibenclamide (ATP-sensitive K(+) channel blocker, 25-100μg/paw), l-arginine (a nitric oxide precursor, 25-100μg/paw) and sodium nitroprusside (125-500μg/paw). The local peripheral ipsilateral, but not contralateral, administration of EA into the right paw (30-300μg/paw) produced a dose-related antinociception during both early and late phases of formalin test which is comparable with morphine (25μg/paw). Moreover, local pre-treatment with l-NAME, methylene blue and glibenclamide dose-dependently prevented EA (100μg/paw)-induced antinociception in late phase. Additionally, administration of l-arginine and sodium nitroprusside significantly potentiated the antinociception induced by EA in the late phase. However, these treatments had no significant effect on antinociceptive response of EA in the early phase of the formalin test. The results of the present study showed that EA-induced local peripheral antinociception during the both phases of formalin test. Also, our data suggested the activation of the l-arginine/NO/cGMP/KATP channels pathway in EA-induced antinociception in late phase of formalin test. Topical application of EA by ointment or jelly might be a useful method to relieving the inflammatory pain states.
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Chloromethane, ≥99.5%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Methylene Blue solution
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%