MilliporeSigma
  • Arg-265: a critical residue of L.donovani cytosolic SHMT in maintaining the binding of THF and catalysis.
Arg-265: a critical residue of L.donovani cytosolic SHMT in maintaining the binding of THF and catalysis.
Experimental parasitology (2014-12-17)
Shashi Gandhi, Neha Gaur, Shagun Krishna, Mohd Imran Siddiqi, Jitendra Kumar Saxena
ABSTRACT
Serine hydroxymethyltransferase belongs to the class of pyridoxal-5-phosphate enzymes along with aspartate aminotransferase. To explore the function of residue(s) involved in binding of the carboxylate group of Tetrahydrofolic acid (THF) to L. donovani cytosolic serine hydroxymethyltransferase (LdcSHMT), the gene was cloned in pET-28(a) vector, overexpressed and purified to homogeneity. With the help of docking results of THF to the active site of protein, the key residues involved in interaction were identified. In an attempt to unravel the function of Arg265 residue involved in binding of the carboxylate group of THF, Arg-265 was mutated to Ala by site-directed mutagenesis. The Arg265Ala-LdcSHMT showed increased Km value (threefold) and decreased kcat/Km value (threefold) for H₄-folate as compared with wild type enzyme. The wild and mutant enzymes exhibited similar Km and kcat/Km values for L-allo-threonine. Unlike the wild type enzyme, mutant failed to form characteristic quinonoid intermediate and was unable to carry out the exchange of α-proton from glycine in the presence of Tetrahydrofolate. These results suggested that Arg265 residue is required for the binding of Tetrahydrofolate and may be the base that abstracts α-proton from glycine, leading to formation of quinonoid intermediate in cytosolic SHMT of L. donovani.
Product Description
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Nitrilotriacetic acid disodium salt, Sigma Grade, ≥99%
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Pyridoxal 5′-phosphate monohydrate, ≥97.0% (NT)
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Serine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Serine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
Tetrahydrofolic acid, ≥65% (when packaged), powder
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, ≥98%
Sigma-Aldrich
Nitrilotriacetic acid trisodium salt, Sigma Grade, ≥98%
Sigma-Aldrich
Pyridoxal 5′-phosphate hydrate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, ≥99.0%, contains 200-400 ppm BHT as inhibitor, ReagentPlus®
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer