MilliporeSigma
  • Pharmacological basis for the medicinal use of Wrightia tinctoria in hypertension and dyslipidemia.
Pharmacological basis for the medicinal use of Wrightia tinctoria in hypertension and dyslipidemia.
Journal of cardiovascular pharmacology (2014-04-01)
Hasan Salman Siddiqi, Amara Majeed, Anwarul Hassan Gilani
ABSTRACT
This study was aimed to offer a possible pharmacological basis regarding the remedial utilization of Wrightia tinctoria in hypertension and dyslipidemia in certain South Asian traditional systems of medicine, using in vivo and in vitro assays. The aqueous methanolic extract of W. tinctoria seeds (Wt.Cr) caused a dose-dependent (1-10 mg/kg) decrease in arterial pressure in anesthetized rats. In the right atria of isolated guinea pigs, Wt.Cr equally inhibited force and rate of spontaneous atrial contractions. When tested on phenylephrine-, high K(+)-, and low K(+)-induced vasoconstrictions in isolated rat aorta, Wt.Cr produced a concentration-dependent vasorelaxation, the most potent being against low K(+)-induced contraction. It also created a rightward shift in the Ca(++) concentration-response curves and suppressed phenylephrine control peaks in a Ca(++)-free environment. In the rat model of tyloxapol-induced dyslipidemia, Wt.Cr produced a decline in the serum levels of total cholesterol and triglycerides. In high fat diet-induced dyslipidemia, it decreased serum total cholesterol and low-density lipoprotein cholesterol, improved high-density lipoprotein cholesterol, and prevented the increase in average body weights by causing a decrease in diet consumption. These data suggest that Wt.Cr(++) lowers blood pressure through a combination of K(+)-channel opening and Ca(++)-channel blocking effects along with antidyslipidemic and weight-reducing properties.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Barium chloride, 99.9% trace metals basis
Sigma-Aldrich
Barium chloride, AnhydroBeads, −10 mesh, 99.95% trace metals basis
Sigma-Aldrich
Barium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Barium chloride, 99.999% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
USP
Verapamil hydrochloride, United States Pharmacopeia (USP) Reference Standard
Phenylephrine, European Pharmacopoeia (EP) Reference Standard
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
4-Aminopyridine, 98%
Sigma-Aldrich
4-Aminopyridine, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
Potassium chloride, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Supelco
ISA (ionic strength adjustment solution: 1 M KCl), 1 M KCl