MilliporeSigma
  • Ammonia-lowering activities and carbamoyl phosphate synthetase 1 (Cps1) induction mechanism of a natural flavonoid.
Ammonia-lowering activities and carbamoyl phosphate synthetase 1 (Cps1) induction mechanism of a natural flavonoid.
Nutrition & metabolism (2015-06-16)
Kazunari Nohara, Youngmin Shin, Noheon Park, Kwon Jeong, Baokun He, Nobuya Koike, Seung-Hee Yoo, Zheng Chen
ABSTRACT
Ammonia detoxification is essential for physiological well-being, and the urea cycle in liver plays a predominant role in ammonia disposal. Nobiletin (NOB), a natural dietary flavonoid, is known to exhibit various physiological efficacies. In the current study, we investigated a potential role of NOB in ammonia control and the underlying cellular mechanism. C57BL/6 mice were fed with regular chow (RC), high-fat (HFD) or high-protein diet (HPD) and treated with either vehicle or NOB. Serum and/or urine levels of ammonia and urea were measured. Liver expression of genes encoding urea cycle enzymes and C/EBP transcription factors was determined over the circadian cycle. Luciferase reporter assays were carried out to investigate function of CCAAT consensus elements on the carbamoyl phosphate synthetase (Cps1) gene promoter. A circadian clock-deficient mouse mutant, Clock (Δ19/Δ19) , was utilized to examine a requisite role of the circadian clock in mediating NOB induction of Cps1. NOB was able to lower serum ammonia levels in mice fed with RC, HFD or HPD. Compared with RC, HFD repressed the mRNA and protein expression of Cps1, encoding the rate-limiting enzyme of the urea cycle. Interestingly, NOB rescued CPS1 protein levels under the HFD condition via induction of the transcription factors C/EBPα and C/EBPβ. Expression of other urea cycle genes was also decreased by HFD relative to RC and again restored by NOB to varying degrees, which, in conjunction with Cps1 promoter reporter analysis, suggested a C/EBP-dependent mechanism for the co-induction of urea cycle genes by NOB. In comparison, HPD markedly increased CPS1 levels relative to RC, yet NOB did not further enrich CPS1 to a significant extent. Using the circadian mouse mutant Clock (Δ19/Δ19) , we also showed that a functional circadian clock, known to modulate C/EBP and CPS1 expression, was required for NOB induction of CPS1 under the HFD condition. NOB, a dietary flavonoid, exhibits a broad activity in ammonia control across varying diets, and regulates urea cycle function via C/EBP-and clock-dependent regulatory mechanisms.
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Nobiletin, ≥97%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Ammonia Assay Kit, sufficient for 100 assays
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, meets USP testing specifications