MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Pharmacological inhibition of p38 MAPK reduces tumor growth in patient-derived xenografts from colon tumors.
Pharmacological inhibition of p38 MAPK reduces tumor growth in patient-derived xenografts from colon tumors.
Oncotarget (2015-04-22)
Jalaj Gupta, Ana Igea, Marilena Papaioannou, Pedro Pablo Lopez-Casas, Elisabet Llonch, Manuel Hidalgo, Vassilis G Gorgoulis, Angel R Nebreda
ABSTRACT
Colorectal cancer is a major health problem and the second cause of cancer related death in western countries. Signaling pathways that control tissue homeostasis are often deregulated during tumorigenesis and contribute to tumor development. Studies in mouse models have shown that the p38 MAPK pathway regulates homeostasis in colon epithelial cells but also plays an important role in colon tumor maintenance. In this study, we have investigated the role of p38 MAPK signaling in patient-derived xenografts (PDXs) from three different human colon tumors representing clinical heterogeneity and that recapitulate the human tumor conditions both at histological and molecular levels. We have found that PH797804, a chemical inhibitor of p38 MAPK, reduces tumor growth of the three PDXs, which correlates with impaired colon tumor cell proliferation and survival. The inhibition of p38 MAPK in PDXs results in downregulation of the IL-6/STAT3 signaling pathway, which is a key regulator of colon tumorigenesis. Our results show the importance of p38 MAPK in human colon tumor growth using a preclinical model, and support that inhibition of p38 MAPK signaling may have therapeutic interest for colon cancer treatment.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
Sigma-Aldrich
Periodic acid, SAJ special grade, ≥98.5%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Periodic acid, 99.999% trace metals basis
Sigma-Aldrich
Periodic acid, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O