MilliporeSigma
  • Development of advanced host cell protein enrichment and detection strategies to enable process relevant spike challenge studies.
Development of advanced host cell protein enrichment and detection strategies to enable process relevant spike challenge studies.
Biotechnology progress (2015-05-28)
Ryan G Soderquist, Mihaela Trumbo, Roger A Hart, Qingchun Zhang, Gregory C Flynn
ABSTRACT
An orthogonal chromatography methodology for the enrichment of host cell protein (HCP) species relative to monoclonal antibody (mAb) products was developed and applied for the successful enrichment of HCP from post-Protein A process pools for seven different mAb products. An advanced two-dimensional liquid chromatography/mass spectrometry platform (2D-LC/MS(E) ) was utilized to demonstrate that the HCP enriched material was representative, in terms of species content, to pre-enriched process pools. The HCP enrichment methodology was scaled up for two different mAb products, and this process relevant enriched HCP material was used to conduct advanced spike challenge studies to demonstrate the utility of the approach for the understanding of (1) quantitative HCP clearance, (2) individual species clearance, and (3) species clearance redundancy across polishing chromatography steps. The combined ability to enrich process relevant HCP, detect individual HCP species with 2D-LC/MS(E) technology, and conduct advanced challenge studies with process relevant material surmounts prior limitations to high integrity process challenge study implementation, and facilitates significant process understanding for development of risk-based control strategies and strategic process design. This also demonstrates implementation of a foundational strategy for conducting spike-challenge studies using process-relevant impurities isolated from processes of interest using orthogonal approaches.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, 1 M
Pricing and availability is not currently available.
SAFC
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Pricing and availability is not currently available.
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, tablet
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Pricing and availability is not currently available.