MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Krill Oil Ameliorates Mitochondrial Dysfunctions in Rats Treated with High-Fat Diet.
Krill Oil Ameliorates Mitochondrial Dysfunctions in Rats Treated with High-Fat Diet.
BioMed research international (2015-08-25)
Alessandra Ferramosca, Annalea Conte, Vincenzo Zara
ABSTRACT
In recent years, several studies focused their attention on the role of dietary fats in the pathogenesis of hepatic steatosis. It has been demonstrated that a high-fat diet is able to induce hyperglycemia, hyperinsulinemia, obesity, and nonalcoholic fatty liver disease. On the other hand, krill oil, a novel dietary supplement of n-3 PUFAs, has the ability to improve lipid and glucose metabolism, exerting possible protective effects against hepatic steatosis. In this study we have investigated the effects of krill oil on mitochondrial energetic metabolism in animals fed a high-fat diet. To this end, male Sprague-Dawley rats were divided into three groups and fed for 4 weeks with a standard diet (control group), a diet with 35% fat (HF group), or a high-fat diet supplemented with 2.5% krill oil (HF+KO group). The obtained results suggest that krill oil promotes the burning of fat excess introduced by the high-fat diet. This effect is obtained by stimulating mitochondrial metabolic pathways such as fatty acid oxidation, Krebs cycle, and respiratory chain complexes activity. Modulation of the expression of carrier proteins involved in mitochondrial uncoupling was also observed. Overall, krill oil counteracts the negative effects of a high-fat diet on mitochondrial energetic metabolism.
Sigma-Aldrich
DL-Malic acid, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
Phospho(enol)pyruvic acid trisodium salt hydrate, ≥97% (enzymatic)
Sigma-Aldrich
Thiamine pyrophosphate, ≥95%
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Succinic acid, BioReagent, BioRenewable, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Succinic acid, ReagentPlus®, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, BioXtra, BioRenewable, ≥99.0%
Sigma-Aldrich
Succinic acid, puriss. p.a., ACS reagent, ≥99.5% (T)
Sigma-Aldrich
DL-Malic acid, ≥98% (capillary GC)
Sigma-Aldrich
DL-Malic acid, 99%
Sigma-Aldrich
DL-Malic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N,N′,N′-Tetramethyl-p-phenylenediamine, 99%, powder
Sigma-Aldrich
Malic acid, meets USP/NF testing specifications
Sigma-Aldrich
Succinic acid, SAJ special grade, ≥99.5%
Sigma-Aldrich
Oxaloacetic acid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97% (HPLC)
Sigma-Aldrich
Oxaloacetic acid, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Oxaloacetic acid, ≥97% (HPLC)
Sigma-Aldrich
Decylubiquinone, ≥97% (HPLC)
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture