MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • All-trans-retinoic acid modulates nitric oxide and interleukin-17A production by peripheral blood mononuclear cells from patients with Alzheimer's disease.
All-trans-retinoic acid modulates nitric oxide and interleukin-17A production by peripheral blood mononuclear cells from patients with Alzheimer's disease.
Neuroimmunomodulation (2015-08-19)
Nassima Behairi, Mourad Belkhelfa, Hamida Mesbah-Amroun, Hayet Rafa, Soreya Belarbi, Meriem Tazir, Chafia Touil-Boukoffa
ABSTRACT
Alzheimer's disease (AD), the most common form of dementia in the elderly, is a neurodegenerative disorder associated with a complex pathophysiology. It is accepted that inflammation contributes to the pathogenesis of AD. All-trans-retinoic acid (ATRA) is a bioactive derivative of vitamin A that has shown immunomodulatory effects in many immune disorders. In our study, we aimed to investigate in vitro immunomodulatory effects of ATRA on inducible nitric oxide synthase (iNOS) expression and interleukin-17A production during AD. Peripheral blood mononuclear cells (PBMCs) isolated from 30 Algerian AD patients and 14 age-matched nondemented controls were treated (or not) with ATRA. Production of NO and IL-17A in culture media was measured by the modified Griess method and enzyme-linked immunosorbent assay, respectively. Expression of iNOS in PBMCs was examined by fluorescence immunostaining. Our results showed higher spontaneous in vitro production of NO related to overexpression of iNOS in AD patients compared to controls. Remarkably, ATRA treatment showed an important downregulatory effect on NO production and iNOS expression in patients. This effect was associated with a reduction in IL-17A production and increased IL-10 release. Taken together, our results indicate that ATRA exerts anti-inflammatory effects in AD. Furthermore, ATRA represents a promising tool for monitoring inflammatory responses associated with disease progression.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)