MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • IL-21 Promotes Pulmonary Fibrosis through the Induction of Profibrotic CD8+ T Cells.
IL-21 Promotes Pulmonary Fibrosis through the Induction of Profibrotic CD8+ T Cells.
Journal of immunology (Baltimore, Md. : 1950) (2015-11-01)
Tia Y Brodeur, Tara E Robidoux, Jason S Weinstein, Joseph Craft, Susan L Swain, Ann Marshak-Rothstein
ABSTRACT
Type 2 effector production of IL-13, a demonstrated requirement in models of fibrosis, is routinely ascribed to CD4(+) Th2 cells. We now demonstrate a major role for CD8(+) T cells in a murine model of sterile lung injury. These pulmonary CD8(+) T cells differentiate into IL-13-producing Tc2 cells and play a major role in a bleomycin-induced model of fibrosis. Differentiation of these Tc2 cells in the lung requires IL-21, and bleomycin treated IL-21- and IL-21R-deficient mice develop inflammation but not fibrosis. Moreover, IL-21R-expressing CD8(+) cells are sufficient to reconstitute the fibrotic response in IL-21R-deficient mice. We further show that the combination of IL-4 and IL-21 skews naive CD8(+) T cells to produce IL-21, which, in turn, acts in an autocrine manner to support robust IL-13 production. Our data reveal a novel pathway involved in the onset and regulation of pulmonary fibrosis and identify Tc2 cells as key mediators of fibrogenesis.
Sigma-Aldrich
Interleukin-2, human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in E. coli, ~10000 U/mL
Sigma-Aldrich
Interleukin-4 from mouse, IL-4, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Interleukin-2 human, recombinant, expressed in Pichia pastoris, suitable for cell culture
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Interleukin-2 human, IL-2, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Interleukin-21 from mouse, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
IL-4 from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, ≥99.7%, suitable for amino acid analysis
Sigma-Aldrich
Acetic acid, 99.5-100.0%
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
Acetic acid solution, 1 N, 1 M
Sigma-Aldrich
Acetic acid, ≥99.7%