Merck
A Novel Angiotensin Type I Receptor Antagonist, Fimasartan, Prevents Doxorubicin-induced Cardiotoxicity in Rats.
Journal of Korean medical science (2015-05-02)
Sung-A Chang, Byung-Kwan Lim, You Jung Lee, Mi-Kyung Hong, Jin-Oh Choi, Eun-Seok Jeon
ABSTRAKT
Angiotensin receptor blockers (ARBs) have organ-protective effects in heart failure and may be also effective in doxorubicin-induced cardiomyopathy (DOX-CMP); however, the efficacy of ARBs on the prevention of DOX-CMP have not been investigated. We performed a preclinical experiment to evaluate the preventive effect of a novel ARB, fimasartan, in DOX-CMP. All animals underwent echocardiography and were randomly assigned into three groups: treated daily with vehicle (DOX-only group, n=22), 5 mg/kg of fimasartan (Low-fima group, n=22), and 10 mg/kg of fimasartan (High-fima group, n=19). DOX was injected once a week for six weeks. Echocardiography and hemodynamic assessment was performed at the 8th week using a miniaturized conductance catheter. Survival rate of the High-fima group was greater (100%) than that of the Low-fima (75%) and DOX-only groups (50%). Echocardiography showed preserved left ventricular (LV) ejection fraction in the High-fima group, but not in the DOX-only group (P=0.002). LV dimensions increased in the DOX-only group; however, remodeling was attenuated in the Low-fima and High-fima groups. Hemodynamic assessment showed higher dP/dt in the High-fima group compared with the DOX-only group. A novel ARB, fimasartan, may prevent DOX-CMP and improve survival rate in a dose-dependent manner in a rat model of DOX-CMP and could be a treatment option for the prevention of DOX-CMP.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)