Merck
PhosFox: a bioinformatics tool for peptide-level processing of LC-MS/MS-based phosphoproteomic data.
Proteome science (2014-07-17)
Sandra Söderholm, Petteri Hintsanen, Tiina Öhman, Tero Aittokallio, Tuula A Nyman
ABSTRAKT
It is possible to identify thousands of phosphopeptides and -proteins in a single experiment with mass spectrometry-based phosphoproteomics. However, a current bottleneck is the downstream data analysis which is often laborious and requires a number of manual steps. Toward automating the analysis steps, we have developed and implemented a software, PhosFox, which enables peptide-level processing of phosphoproteomic data generated by multiple protein identification search algorithms, including Mascot, Sequest, and Paragon, as well as cross-comparison of their identification results. The software supports both qualitative and quantitative phosphoproteomics studies, as well as multiple between-group comparisons. Importantly, PhosFox detects uniquely phosphorylated peptides and proteins in one sample compared to another. It also distinguishes differences in phosphorylation sites between phosphorylated proteins in different samples. Using two case study examples, a qualitative phosphoproteome dataset from human keratinocytes and a quantitative phosphoproteome dataset from rat kidney inner medulla, we demonstrate here how PhosFox facilitates an efficient and in-depth phosphoproteome data analysis. PhosFox was implemented in the Perl programming language and it can be run on most common operating systems. Due to its flexible interface and open source distribution, the users can easily incorporate the program into their MS data analysis workflows and extend the program with new features. PhosFox source code, implementation and user instructions are freely available from https://bitbucket.org/phintsan/phosfox. PhosFox facilitates efficient and more in-depth comparisons between phosphoproteins in case-control settings. The open source implementation is easily extendable to accommodate additional features for widespread application use cases.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, tablet
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride solution, 5 M
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Informacje o cenach i dostępności nie są obecnie dostępne.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Informacje o cenach i dostępności nie są obecnie dostępne.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Informacje o cenach i dostępności nie są obecnie dostępne.