Merck
Determination of Synacthen(®) in dried blood spots for doping control analysis using liquid chromatography tandem mass spectrometry.
Analytical and bioanalytical chemistry (2015-04-13)
Laura Tretzel, Andreas Thomas, Hans Geyer, Philippe Delahaut, Wilhelm Schänzer, Mario Thevis
ABSTRAKT
Dried blood spot (DBS) sampling, a technique used for taking whole blood samples dried on a filter paper, was initially reported in 1963 by Robert Guthrie. While the diagnostic analysis of metabolic disorders in newborns was the focus of investigations at that time, the number of established applications for preclinical drug development, toxicological studies, and therapeutic drug monitoring increased enormously in the last decades. As a consequence of speed, simplicity, and minimal invasiveness, DBS recommends itself as the preferential technique in sports drug testing. The present approach highlights for the first time the development of a screening assay for the analysis of the synthetic human adrenocorticotropic hormone tetracosactide hexaacetate (Synacthen(®)) in DBS using liquid chromatography tandem mass spectrometry. Highly purified sample extracts were obtained by an advanced sample preparation procedure including the addition of an internal standard (d8-tetracosactide) and immunoaffinity purification. The method's overall recovery was 27.6 %, and the assay's imprecision was calculated between 8.1 and 17.9 % for intraday and 12.9 to 20.5 % for interday measurements. Stability of the synthetic peptide in DBS was shown for at least 10 days at room temperature and presents a major benefit, since a rapid degradation in conventionally applied matrices such as urine or plasma is well known. With a limit of detection of 50 pg/mL, a detection window of several hours is expected considering reported steady-state plasma levels of 300 pg/mL after intramuscular application of Synacthen(®) Depot (1 mg). The analysis of authentic DBS samples within the scope of an administration study with 250 μg Synacthen(®) (short stimulation test) demonstrated the great potential of the developed assay to simplify the analysis of Synacthen(®) for doping control purposes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, suitable for electrophoresis, SDS-PAGE tested
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, ≥98% (NMR)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Supelco
Tris(2-carboxyethyl)phosphine hydrochloride solution, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%