MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Open-tubular CEC with a new triethanolamine bonded stationary phase for biomolecule separation.
Open-tubular CEC with a new triethanolamine bonded stationary phase for biomolecule separation.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-09)
Cemil Aydoğan
ABSTRACT
This study describes the preparation and electrochromatographic application of a new open-tubular capillary column with triethanolamine functionalized stationary phase. The stationary phase was synthesized by in situ grafting polymerization with 3-chloro-2-hydroxypropylmethacrylate based reactive monomer, and followed triethanolamine functionalization. The different 3-chloro-2-hydroxypropylmethacrylate contents on the separation efficiency were studied. The results indicated that the 3-chloro-2-hydroxypropylmethacrylate content (e.g., 15.3v/v%) on the inner surface of the capillary was very important for final preparation of the polymer stationary phase. The electrochromatographic characterization of the stationary phase was performed using alkylbenzene derivatives. The pH effect on the electroosmotic flow was also investigated. The open tubular column functionalized with triethanolamine allowed to operate in the anodic electroosmotic flow mode in the system. With anodic electroosmotic flow mode, favorable separations of the amino acids and the nucleosides were successfully achieved with high column efficiens ranging from 142000 to 257000 plates/m. Good repeatability was gained with relative standard deviation of the migration time and peak areas less than 2.2% for run to run (n=5) and less than 3.2% day to day (n=3). Furthermore, real sample applicability of this column to the separation of amino acids in white tea sample was demonstrated.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Cytidine, BioReagent, suitable for cell culture, powder, ≥99%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymidine, ≥99%
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%