MilliporeSigma
  • Boosting D-lactate production in engineered cyanobacteria using sterilized anaerobic digestion effluents.
Boosting D-lactate production in engineered cyanobacteria using sterilized anaerobic digestion effluents.
Bioresource technology (2014-08-02)
Whitney D Hollinshead, Arul M Varman, Le You, Zachary Hembree, Yinjie J Tang
ABSTRACT
Anaerobic digestion (AD) is an environmentally friendly approach to waste treatment, which can generate N and P-rich effluents that can be used as nutrient sources for microalgal cultivations. Modifications of AD processes to inhibit methanogenesis leads to the accumulation of acetic acid, a carbon source that can promote microalgal biosynthesis. This study tested different AD effluents from municipal wastes on their effect on D-lactate production by an engineered Synechocystis sp. PCC 6803 (carrying a novel lactate dehydrogenase). The results indicate that: (1) AD effluents can be supplemented into the modified BG-11 culture medium (up to 1:4 volume ratio) to reduce N and P cost; (2) acetate-rich AD effluents enhance D-lactate synthesis by ∼ 40% (1.2g/L of D-lactate in 20 days); and (3) neutral or acidic medium had a deleterious effect on lactate secretion and biomass growth by the engineered strain. This study demonstrates the advantages and guidelines in employing wastewater for photomixotrophic biosynthesis using engineered microalgae.
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Chlorophyll a, from Anacystis nidulans algae
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, ≥95%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Chlorophyll a, from spinach
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, SAJ first grade, ≥99.0%
Sigma-Aldrich
Tetrahydrofuran, JIS special grade, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, ≥99.0%, contains 200-400 ppm BHT as inhibitor, ReagentPlus®
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, contains no stabilizer
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Supelco
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, LiChropur, ≥95.0% (GC)
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C