MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Cdc42 inhibitor ML141 enhances G-CSF-induced hematopoietic stem and progenitor cell mobilization.
Cdc42 inhibitor ML141 enhances G-CSF-induced hematopoietic stem and progenitor cell mobilization.
International journal of hematology (2014-10-16)
Chong Chen, Xuguang Song, Sha Ma, Xue Wang, Jie Xu, Huanxin Zhang, Qingyun Wu, Kai Zhao, Jiang Cao, Jianlin Qiao, Xiaoshen Sun, Depeng Li, Lingyu Zeng, Zhengyu Li, Kailin Xu
ABSTRACT
G-CSF is the most often used agent in clinical hematopoietic stem and progenitor cell (HSPC) mobilization. However, in about 10 % of patients, G-CSF does not efficiently mobilize HSPC in clinically sufficient amounts. Cdc42 activity is involved in HSPC mobilization. In the present study, we explore the impact of Cdc42 inhibitor ML141 on G-CSF-mediated HSPC mobilization in mice. We found that the use of ML141 alone only triggered modest HSPC mobilization effect in mice. However, combination of G-CSF and ML141 significantly promoted HPSC counts and colony forming units in peripheral blood, as compared to mice treated with G-CSF alone. ML141 did not significantly alter the levels of SDF-1 and MMP-9 in the bone marrow, when used alone or in combination with G-CSF. We also found that G-CSF administration significantly increases the level of GTP-bound Cdc42, but does not alter the expression of Cdc42 in the bone marrow. Our data indicate that the Cdc42 signal is a negative regulator in G-CSF-mediated HSPC mobilization, and that inhibition of the Cdc42 signal efficiently improves mobilization efficiency. These findings may provide a new strategy for efficient HSPC mobilization, especially in patients with poor G-CSF response.
Product Description
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium fluoride, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water