MilliporeSigma
  • Inhibitory effect of Careya arborea on inflammatory biomarkers in carrageenan-induced inflammation.
Inhibitory effect of Careya arborea on inflammatory biomarkers in carrageenan-induced inflammation.
Pharmaceutical biology (2014-12-05)
Rayhana Begum, Manju Sharma, K K Pillai, Vidhu Aeri, Manjur Ali Sheliya
ABSTRACT
Careya arborea Roxb. (Lecythidaceae) has multiple applications in traditional medicine; it exhibits analgesic, antibacterial, anti-inflammatory, antiulcer, and protective effects. However, the effect of C. arborea on biochemical and immmunological inflammatory mediators has not been explored. The present study investigates the anti-inflammatory potential of the methanol extract of C. arborea stem bark and further assesses its possible mechanism on the modulation of inflammatory biomarkers. Anti-inflammatory activity of C. arborea methanol extract (CAME) was evaluated (100 and 200 mg/kg, p.o) using indomethacin (10 mg/kg, p.o) as the standard drug in Wistar albino rats. Inflammation was induced by injecting 0.1 ml carrageenan (1% w/v) into the left hind paw. The anti-inflammatory mechanism was studied by measuring malondialdehyde (MDA), C-reactive protein (CRP), nitric oxide (NO), myeloperoxidase (MPO), TNF-α, and IL-1β levels in both control and treated groups. A protocol has also been established to quantify quercetin and betulinic acid content in CAME using HPTLC fingerprint. Careya arborea significantly (p < 0.001) decreased carrageenan-induced paw edema, showed a reduction of 48.87 and 65.53% at doses of 100 and 200 mg/kg, respectively. Moreover, CAME significantly decreased the MDA, CRP, NO, and MPO levels, elevated by carrageenan induced inflammation. CAME also markedly down-regulated serum TNF-α and IL-1β levels. These findings were further supported by the histological study. The content of quercetin and betulinic acid in CAME was found to be 0.177 and 3.14%, respectively. Several mechanisms, including the inhibition of pro-inflammatory cytokines, enzymes and mediators release, appear to account for the anti-inflammatory potential of C. arborea.
Sigma-Aldrich
Hydrogen peroxide solution, SAJ first grade, ≥30.0%
Pricing and availability is not currently available.
Supelco
Sulfanilamide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Pricing and availability is not currently available.
USP
Sulfanilamide Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Pricing and availability is not currently available.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Pricing and availability is not currently available.
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Pricing and availability is not currently available.
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Betulinic acid, technical grade, 90%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfanilamide, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Pricing and availability is not currently available.
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Pricing and availability is not currently available.
Sigma-Aldrich
Betulinic acid, ≥98% (HPLC)
Pricing and availability is not currently available.
Supelco
Betulinic acid, analytical standard
Pricing and availability is not currently available.
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Pricing and availability is not currently available.