MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Regulation of Cigarette Smoke Induction of IL-8 in Macrophages by AMP-activated Protein Kinase Signaling.
Regulation of Cigarette Smoke Induction of IL-8 in Macrophages by AMP-activated Protein Kinase Signaling.
Journal of cellular physiology (2014-12-17)
Hsin-Kuo Ko, Hung-Fu Lee, An-Hsuan Lin, Meng-Han Liu, Ching-I Liu, Tzong-Shyuan Lee, Yu Ru Kou
ABSTRACT
Inhaled cigarette smoke (CS) causes persistent lung inflammation in smokers. Interleukin 8 (IL-8) released from macrophages is a key chemokine during initiation and progression of CS-induced lung inflammation, yet its regulation is largely unknown. AMP-activated protein kinase (AMPK), a crucial energy homeostasis regulator, may modulate inflammation. Here we report that CS extract (CSE) increased the level of intracellular reactive oxygen species (ROS), activating AMPK, mitogen-activated protein kinases (MAPKs), and NF-κB, as well as inducing IL-8, in human macrophages. N-acetyl-cysteine (ROS scavenger) or hexamethonium [nicotinic acetylcholine receptor (nAChR) antagonist] attenuated the CSE-induced increase in intracellular ROS, activation of AMPK and NF-κB, as well as IL-8 induction, which suggests that nAChRs and ROS are important. Prevention of AMPK activation by compound C or AMPK siRNA reduced CSE-induced IL-8 production, confirming the role of AMPK. Compound C or AMPK siRNA also inhibited the activation of MAPKs and NF-κB by CSE, which suggests that these molecules are downstream of AMPK. Additionally, exposure of human macrophages to nicotine activated AMPK and induced IL-8 and that these effects were lessened by hexamethonium or compound C, implying that nicotine in CS may be causative. Furthermore, chronic CS exposure in mice promoted AMPK phosphorylation and expression of MIP-2 (an IL-8 homolog) in infiltrated macrophages and in lung tissues, as well as induced lung inflammation, all of which were reduced by compound C treatment. Thus, we identified a novel nAChRs-dependent, ROS-sensitive, AMPK/MAPKs/NF-κB signaling pathway, which seems to be important to CS-induced macrophage IL-8 production and possibly to lung inflammation.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Hexamethonium bromide
Sigma-Aldrich
(±)-Nicotine, ≥99% (TLC), liquid
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
N-Acetyl-L-cysteine, SAJ special grade, 98.0-102.0%