MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Hypoxia-inducible TAp73 supports tumorigenesis by regulating the angiogenic transcriptome.
Hypoxia-inducible TAp73 supports tumorigenesis by regulating the angiogenic transcriptome.
Nature cell biology (2015-03-17)
Iqbal Dulloo, Beng Hooi Phang, Rashidah Othman, Soo Yong Tan, Aadhitthya Vijayaraghavan, Liang Kee Goh, Marta Martin-Lopez, Margarita M Marques, Chun Wei Li, De Yun Wang, Maria Carmen Marín, Wa Xian, Frank McKeon, Kanaga Sabapathy
ABSTRACT
The functional significance of the overexpression of unmutated TAp73, a homologue of the tumour suppressor p53, in multiple human cancers is unclear, but raises the possibility of unidentified roles in promoting tumorigenesis. We show here that TAp73 is stabilized by hypoxia, a condition highly prevalent in tumours, through HIF-1α-mediated repression of the ubiquitin ligase Siah1, which targets TAp73 for degradation. Consequently, TAp73-deficient tumours are less vascular and reduced in size, and conversely, TAp73 overexpression leads to increased vasculature. Moreover, we show that TAp73 is a critical regulator of the angiogenic transcriptome and is sufficient to directly activate the expression of several angiogenic genes.  Finally, expression of TAp73 positively correlates with these angiogenic genes in several human tumours, and the angiogenic gene signature is sufficient to segregate the TAp73(Hi)- from TAp73(Low)-expressing tumours. These data demonstrate a pro-angiogenic role for TAp73 in supporting tumorigenesis, providing a rationale for its overexpression in cancers.
Sigma-Aldrich
DMOG, ≥98% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Ethylmaleimide, purum p.a., ≥99.0% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, BioUltra, ≥99.0% (HPLC)
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Imidazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)