Merck
Coculture with mesenchymal stem cells results in improved viability and function of human hepatocytes.
Cell transplantation (2013-10-23)
Emer Fitzpatrick, Yue Wu, Paramjeet Dhadda, Robin D Hughes, Ragai R Mitry, Hong Qin, Sharon C Lehec, Nigel D Heaton, Anil Dhawan
ABSTRAKT
Hepatocyte transplantation is becoming an accepted therapy for acute liver failure, either as a bridge to liver regeneration or to organ transplantation. Hepatocytes provide liver function in place of the failing organ. The maintenance of sufficient viability and function of the transplanted hepatocytes is a concern. There is a lot of recent interest in mesenchymal stem cells (MSCs) for the provision of structural and trophic support to hepatocytes, but few studies currently use primary human hepatocytes. The aim of this study was to investigate if coculture of human MSCs with cryopreserved human hepatocytes may improve their function and viability, thus with potential for cellular therapy of liver disease. MSCs were isolated from human umbilical cord or adipose tissue. Hepatocytes were isolated from donor organs unsuitable for transplantation. MSCs and hepatocytes were cocultured in both direct and indirect contact. Conditioned medium (CM) from cocultured MSCs and hepatocytes was also used on hepatocytes. Viability and liver-specific function were compared between test and controls. Human hepatocytes that were cocultured directly with MSCs demonstrated improved production of albumin from day 5 to day 25 of culture. This effect was most prominent at day 15. Likewise, urea production was improved in coculture from day 5 to 25. Indirect coculture demonstrated improved albumin production by day 4 (1,107 ng/ml) versus hepatocyte monoculture (940 ng/ml). Hepatocytes in CM demonstrated a nonsignificant improvement in function. The viability of cocultured hepatocytes was superior to that of monocultured cells with up to a 16% improvement. Thus, coculture of human hepatocytes with MSCs demonstrates both improved function and viability. The effect is seen mainly with direct coculture but can also be seen in indirect culture and with CM. Such coculture conditions may convey major advantages in hepatocyte survival and function for cell transplantation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
USP
Kwas askorbinowy, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma