Merck
  • Rapid shedding of proinflammatory microparticles by human mononuclear cells exposed to cigarette smoke is dependent on Ca2+ mobilization.
Rapid shedding of proinflammatory microparticles by human mononuclear cells exposed to cigarette smoke is dependent on Ca2+ mobilization.
Inflammation research : official journal of the European Histamine Research Society ... [et al.] (2014-03-07)
Cinzia Cordazzo, Silvia Petrini, Tommaso Neri, Stefania Lombardi, Yuri Carmazzi, Roberto Pedrinelli, Pierluigi Paggiaro, Alessandro Celi
ABSTRAKT
Microparticles are membrane vesicles shed by cells upon activation and apoptosis. Agonists capable of inducing microparticle generation include cytokines, bacterial products, P-selectin, histamine. Cigarette smoke extract has also been recognized as an agonist involved in microparticle generation with an apoptosis-dependent mechanism. We investigated the possibility that cigarette smoke extract induces the rapid generation of proinflammatory microparticles by human mononuclear cells with a calcium-dependent mechanism. Human mononuclear cells were exposed to cigarette smoke extract. [Ca(2+)]i mobilization was assessed with the fluorescent probe Fluo-4 NW. Microparticles were quantified with a prothrombinase assay and by flow cytometry. Normal human bronchial epithelial cells and A549 alveolar cells were incubated with cigarette smoke extract-induced microparticles and the generation of ICAM-1, IL-8, and MCP-1 was assessed by ELISA. Exposure to cigarette smoke extract induced a rapid increase in [Ca(2+)]i mobilization. Microparticle generation was also increased. EGTA, verapamil and the calmodulin inhibitor, W-7, inhibited microparticle generation. Incubation of lung epithelial cells with cigarette smoke extract-induced microparticles increased the expression of proinflammatory mediators. Exposure of mononuclear cells to cigarette smoke extract causes a rapid shedding of microparticles with a proinflammatory potential that might add to the mechanisms of disease from tobacco use.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Calcium Ionophore A23187, ≥98% (TLC), powder
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma