Merck
  • Molecular characterization and hypoxia-induced upregulation of neuronal nitric oxide synthase in Atlantic croaker: Reversal by antioxidant and estrogen treatments.
Molecular characterization and hypoxia-induced upregulation of neuronal nitric oxide synthase in Atlantic croaker: Reversal by antioxidant and estrogen treatments.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology (2015-04-07)
Md Saydur Rahman, Peter Thomas
ABSTRAKT
Neuronal nitric oxide synthase (nNOS) catalyzes production of nitric oxide in vertebrate brains. Recent findings indicate that endothelial NOS and reactive oxygen species (ROS) are significantly increased during hypoxic stress and are modulated by antioxidants. However, the influence of antioxidants and steroids on nNOS upregulation by hypoxia is largely unknown. In this study, we characterized nNOS cDNA and examined the effects of hypoxia and antioxidant and steroid treatments on nNOS expression in Atlantic croaker hypothalamus. Hypoxia exposure (dissolved oxygen, DO: 1.7 mg/L for 2 and/or 4weeks) caused significant increases in hypothalamic nNOS mRNA, protein and its neuronal expression. Hypothalamic nNOS expression and superoxide radical (O2(·-), an index of ROS) production were increased by pharmacological treatment of fish exposed to normoxic conditions with N-ethylmaleimide, an alkene drug which covalently modifies sulfhydryl groups and inhibits aromatase activity. In contrast, treatments with Nω-nitro-L-arginine methyl ester, a competitive NOS-inhibitor, or vitamin E, an antioxidant, prevented the upregulation of O2(·-) production and nNOS expression in hypoxia-exposed (DO: 1.7 mg/L for 4 weeks) fish. Moreover, treatment with 1,4,6-androstatrien-3,17-dione, an aromatase inhibitor, increased hypothalamic O2(·-) production and nNOS expression in normoxic control fish; whereas estradiol-17β treatment significantly reduced O2(·-) production and nNOS expression in hypoxia-exposed fish. Double-labeled immunohistochemical results showed that nNOS and aromatase proteins are co-expressed in the hypothalamus. Taken together, the results suggest that upregulation of nNOS and ROS in the croaker hypothalamus in response to hypoxia is influenced by antioxidant and overall estrogen status.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
SAFC
MOPS
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%