MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • PX-12 induces apoptosis in Calu-6 cells in an oxidative stress-dependent manner.
PX-12 induces apoptosis in Calu-6 cells in an oxidative stress-dependent manner.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-11-14)
Bo Ra You, Hye Rim Shin, Bo Ram Han, Woo Hyun Park
ABSTRACT
PX-12 (1-methylpropyl 2-imidazolyl disulfide) as a thioredoxin (Trx) inhibitor has an anti-tumor effect. However, there is no report about the toxicological effect of PX-12 on lung cancer cells. Here, we investigated the anti-growth effects of PX-12 on Calu-6 lung cancer cells in relation to reactive oxygen species (ROS) and glutathione (GSH) levels. PX-12 induced the growth inhibition of Calu-6 cells with IC50 of nearly 3 μM at 72 h. In contrast, PX-12 did not affect the growth of human small airway epithelial cells (HSAECs). Cell cycle distribution analysis indicated that PX-12 significantly induced a G2/M phase arrest in Calu-6 cells. PX-12 also increased the number of annexin V-FITC-positive cells in Calu-6 cells. All the tested caspase inhibitors markedly prevented Calu-6 cell death induced by PX-12. With regard to ROS and GSH levels, PX-12 increased ROS levels containing O2(·-) in Calu-6 cells and induced the depletion of GSH. N-acetyl cysteine (NAC), which is a well-known antioxidant, significantly reduced O2(·-) level in PX-12-treated Calu-6 cells and prevented apoptosis and GSH depletion in these cells. In conclusion, it is the first report that PX-12 inhibited the growth of Calu-6 cells via a G2/M phase arrest as well as apoptosis, which effect was related to the intracellular increases in ROS levels.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Z-Ile-Glu(O-ME)-Thr-Asp(O-Me) fluoromethyl ketone, ≥90% (TLC), powder
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Rhodamine 123, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis