MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Protection of differentiated neuronal NG108-15 cells from P2X7 receptor-mediated toxicity by taurine.
Protection of differentiated neuronal NG108-15 cells from P2X7 receptor-mediated toxicity by taurine.
Pharmacological reports : PR (2014-06-21)
Chia-Chia Chao, Paul Chan, Chang-Shin Kuo, Chi-Li Gong, Tzu-Hurng Cheng, Zhong-Min Liu, Pei-Chen Shen, Chieh-Chen Huang, Yuk-Man Leung
ABSTRACT
Strong P2X7 receptor (P2X7R) activation causes Ca(2+) overload and consequent cell death. We previously showed that depletion of Ca(2+) stores and endoplasmic reticulum (ER) stress in differentiated NG108-15 neuronal cells contributed to P2X7R-mediated cytotoxicity. In this work, we assessed whether taurine (2-aminoethanesulfonic acid) could prevent this P2X7R-mediated cytotoxicity in this neuronal cell line. Cytotoxicity markers were assessed by MTT assay and Western blotting. Cytosolic Ca(2+) and mitochondrial Ca(2+) concentrations were measured microfluorimetrically using fura-2 and rhod-2, respectively. Intracellular reactive oxygen species (ROS) production was assayed by the indicator 2',7'-dichlorodihydrofluorescein diacetate. Selective P2X7R agonist BzATP treatment causes neuronal cell death by causing cytosolic Ca(2+) overload, depletion of Ca(2+) stores, endoplasmic reticulum (ER) stress, and caspase-3 activation (cleaved caspase 3). Remarkably, taurine (10mM) pretreatment could prevent P2X7R-mediated neuronal cell death by blocking BzATP-mediated ER stress as determined by phosphorylated eukaryotic translation initiation factor 2α (peIF2α) and C/EBP-homologous protein (CHOP). However, taurine did not block BzATP-induced Ca(2+) overload and depletion of ER Ca(2+) stores. Interestingly, P2X7R activation did not result in mitochondrial Ca(2+) overload, nor did it affect mitochondrial membrane potential. BzATP-induced generation of intracellular reactive oxygen species (ROS) was prevented by taurine. The neuroprotective effect by taurine is attributed to the suppression of P2X7R-mediated ER stress and ROS formation.
Product Description
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Taurine, United States Pharmacopeia (USP) Reference Standard
Ouabain, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Taurine, ≥98%, FG
Sigma-Aldrich
Xestospongin C, film
Sigma-Aldrich
Cyclopiazonic acid from Penicillium cyclopium, ≥98% (HPLC), powder
SAFC
Taurine
Sigma-Aldrich
Rhodamine 123, mitochondrial specific fluorescent dye
Sigma-Aldrich
Rhodamine 123, BioReagent, suitable for fluorescence, ≥85% (HPLC)
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Sigma-Aldrich
Digitonin, Used as non-ionic detergent
Sigma-Aldrich
Digitonin, ~50% (TLC)
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Supelco
Forskolin, analytical standard
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (HPLC), powder
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture