MilliporeSigma
  • Harmonization of QSAR Best Practices and Molecular Docking Provides an Efficient Virtual Screening Tool for Discovering New G-Quadruplex Ligands.
Harmonization of QSAR Best Practices and Molecular Docking Provides an Efficient Virtual Screening Tool for Discovering New G-Quadruplex Ligands.
Journal of chemical information and modeling (2015-09-12)
Daimel Castillo-González, Jean-Louis Mergny, Aurore De Rache, Gisselle Pérez-Machado, Miguel Angel Cabrera-Pérez, Orazio Nicolotti, Antonellina Introcaso, Giuseppe Felice Mangiatordi, Aurore Guédin, Anne Bourdoncle, Teresa Garrigues, Federico Pallardó, M Natália D S Cordeiro, Cesar Paz-Y-Miño, Eduardo Tejera, Fernanda Borges, Maykel Cruz-Monteagudo
ABSTRACT
Telomeres and telomerase are key players in tumorogenesis. Among the various strategies proposed for telomerase inhibition or telomere uncapping, the stabilization of telomeric G-quadruplex (G4) structures is a very promising one. Additionally, G4 stabilizing ligands also act over tumors mediated by the alternative elongation of telomeres. Accordingly, the discovery of novel compounds able to act on telomeres and/or inhibit the telomerase enzyme by stabilizing DNA telomeric G4 structures as well as the development of approaches efficiently prioritizing such compounds constitute active areas of research in computational medicinal chemistry and anticancer drug discovery. In this direction, we applied a virtual screening strategy based on the rigorous application of QSAR best practices and its harmonized integration with structure-based methods. More than 600,000 compounds from commercial databases were screened, the first 99 compounds were prioritized, and 21 commercially available and structurally diverse candidates were purchased and submitted to experimental assays. Such strategy proved to be highly efficient in the prioritization of G4 stabilizer hits, with a hit rate of 23.5%. The best G4 stabilizer hit found exhibited a shift in melting temperature from FRET assay of +7.3 °C at 5 μM, while three other candidates also exhibited a promising stabilizing profile. The two most promising candidates also exhibited a good telomerase inhibitory ability and a mild inhibition of HeLa cells growth. None of these candidates showed antiproliferative effects in normal fibroblasts. Finally, the proposed virtual screening strategy proved to be a practical and reliable tool for the discovery of novel G4 ligands which can be used as starting points of further optimization campaigns.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, SAJ special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate hydrate, 98%
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)