MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • PLA-PEG Coated Multifunctional Imaging Probe for Targeted Drug Delivery.
PLA-PEG Coated Multifunctional Imaging Probe for Targeted Drug Delivery.
Molecular pharmaceutics (2015-04-15)
Hongyu Chen, Susan He
ABSTRACT
Monitoring of drugs in vivo has been recognized as an urgent need for successful chemotherapy treatment. However, to find an appropriate tool for this purpose has been proved as a great challenge. Here, we report a PLA-PEG coated hollow X-ray and upconversion nanophosphor with folic acid as a tumor targeting ligand for a novel drug delivery system. In contrast with previously reported PLA-PEG based or PLA-PEG coated inorganic nanoparticles, our X-ray and upconversion nanophosphor based drug carriers can be used simultaneously as the drug delivery carrier and also serve as labels to track the loaded drugs (paclitaxel) by optical luminescence excited by X-ray, near-infrared light and magnetic resonance imaging. In addition, the hollow space created in our drug carriers dramatically increases the amount of loaded drug. Besides, folic acid on the surface of the drug loaded nanophosphors enhanced cellular uptake and significantly increased the drug efficacy compared with the free drug. We anticipate that these multifunctional nanocarriers will result in significant improvement and serve as a platform in the field of drug delivery.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Toluene, JIS 1000, suitable for residue analysis, ≥99.8%
Sigma-Aldrich
Toluene, SAJ first grade, ≥99.0%
Sigma-Aldrich
(3S)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione, 98%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Toluene, HPLC grade, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Gadolinium, chips
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Gadolinium, −40 mesh, 99% trace rare earth metals basis
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Toluene, suitable for HPLC, ≥99.9%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Sigma-Aldrich
Sulfur, powder, 99.98% trace metals basis