MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Genetic characterization of epizootic hemorrhagic disease virus strains isolated from cattle in Israel.
Genetic characterization of epizootic hemorrhagic disease virus strains isolated from cattle in Israel.
The Journal of general virology (2015-02-24)
William C Wilson, Mark G Ruder, Eyal Klement, Dane C Jasperson, Hagai Yadin, David E Stallknecht, Daniel G Mead, Elizabeth Howerth
ABSTRACT
Epizootic hemorrhagic disease virus (EHDV), a member of the genus Orbivirus not reported previously in Israel, was isolated from Israeli cattle during a 'bluetongue-like' disease outbreak in 2006. To ascertain the origin of this new virus, three isolates from the outbreak were fully sequenced and compared with available sequences. Whilst the L2 gene segment clustered with the Australian EHDV serotype 7 (EHDV-7) reference strain, most of the other segments were clustered with EHDV isolates of African/Middle East origin, specifically Bahrain, Nigeria and South Africa. The M6 gene had genetic relatedness to the Australian/Asian strains, but with the limited data available the significance of this relationship is unclear. Only one EHDV-7 L2 sequence was available, and as this gene encodes the serotype-specific epitope, the relationship of these EHDV-7 L2 genes to an Australian EHDV-7 reflects the serotype association, not necessarily the origin. The genetic data indicated that the strains affecting Israel in 2006 may have been related to similar outbreaks that occurred in North Africa in the same year. This finding also supports the hypothesis that EHDV entered Israel during 2006 and was not present there before this outbreak.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 0.2 M
Sigma-Aldrich
Sodium hydroxide, SAJ first grade, ≥95.0%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)